N'-(Furan-2-ylmethylene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46909-1G 46909-5G 46909-25G
77341-756EA 104.03 USD
77341-756 77363-582 77375-120
N'-(Furan-2-ylmethylene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(Furan-2-ylmethylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃S
MW: 264.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14859984
CAS Number: 1820750-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR