1,4-Diphenylnaphthalene ≥95%

Supplier: AOBChem USA

Warning

46907-1G 46907-5G 46907-25G
77341-964EA 109.18 USD
77341-964 77363-578 77375-118
1,4-Diphenylnaphthalene ≥95%
1,4-Diphenylnaphthalene

Precaución: for Research Use Only

Formula: C₂₂H₁₆
Storage Temperature: Ambient
MDL Number: MFCD32191355
CAS Number: 796-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR