2-(3-Chloro-2,4-difluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

46905-1G 46905-5G 46905-25G
77340-682EA 60.77 USD
77340-682 77363-576 77375-116
2-(3-Chloro-2,4-difluorophenyl)-1,3-dioxolane ≥95%
2-(3-Chloro-2,4-difluorophenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O₂
MW: 220.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707212
CAS Number: 2432848-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR