1-(2-Chloro-3-fluoropyridin-4-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

46890-500MG 46890-1G 46890-5G 46890-25G
77327-268EA 210.12 USD
77327-268 77350-840 77363-570 77386-150
1-(2-Chloro-3-fluoropyridin-4-yl)ethanol ≥97%
1-(2-Chloro-3-fluoropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇ClFNO
Storage Temperature: Ambient
MDL Number: MFCD32707211
CAS Number: 2432848-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR