Ethyl 5-ethoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

46889-500MG 46889-1G 46889-25G 46889-5G
77327-266EA 285.31 USD
77327-266 77350-838 77386-148 77375-110
Ethyl 5-ethoxy-2-methylbenzoate ≥97%
Ethyl 5-ethoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403842
CAS Number: 2624417-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR