Methyl 5-ethoxy-2-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
46888-500MG
46888-1G
46888-5G
46888-25G
77327-264EA
266.77
USD
77327-264
77350-836
77363-568
77386-146
Methyl 5-ethoxy-2-methylbenzoate ≥97%
Methyl 5-ethoxy-2-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₁H₁₄O₃ MW: 194.227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29054759 CAS Number: 104216-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...