4-Ethoxy-2-(methoxymethyl)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

46887-500MG 46887-1G 46887-5G 46887-25G
77327-262EA 222.48 USD
77327-262 77350-834 77363-566 77386-144
4-Ethoxy-2-(methoxymethyl)-1-methylbenzene ≥97%
4-Ethoxy-2-(methoxymethyl)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707210
CAS Number: 2432849-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR