(5-Isopropoxy-2-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

46884-500MG 46884-1G 46884-5G 46884-25G
77327-260EA 208.06 USD
77327-260 77350-832 77363-564 77386-142
(5-Isopropoxy-2-methylphenyl)methanol ≥95%
(5-Isopropoxy-2-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756326
CAS Number: 2764728-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR