Methyl 5-isopropoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

46883-500MG 46883-1G 46883-5G 46883-25G
77327-258EA 163.77 USD
77327-258 77346-114 77363-562 77386-140
Methyl 5-isopropoxy-2-methylbenzoate ≥97%
Methyl 5-isopropoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD23115028
CAS Number: 1394932-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR