Ethyl 5-isopropoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

46882-500MG 46882-1G 46882-5G 46882-25G
77327-256EA 163.77 USD
77327-256 77346-112 77363-560 77386-138
Ethyl 5-isopropoxy-2-methylbenzoate ≥97%
Ethyl 5-isopropoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902701
CAS Number: 2586127-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR