Ethyl 5-bromo-4-fluoro-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

46877-1G 46877-5G 46877-25G
77342-120EA 114.33 USD
77342-120 77363-558 77375-108
Ethyl 5-bromo-4-fluoro-2-methoxybenzoate ≥97%
Ethyl 5-bromo-4-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD30717204
CAS Number: 2114116-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR