1-Bromo-4-iodo-5-methoxy-2-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46869-500MG 46869-1G 46869-5G 46869-25G
77327-254EA 178.19 USD
77327-254 77346-646 77363-556 77386-136
1-Bromo-4-iodo-5-methoxy-2-(trifluoromethyl)benzene ≥95%
1-Bromo-4-iodo-5-methoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₅BrF₃IO
MW: 380.928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707208
CAS Number: 2384872-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR