3-(tert-butoxy)-5-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

46864-500MG 46864-1G 46864-25G 46864-5G
77327-252EA 284.28 USD
77327-252 77350-830 77386-134 77375-104
3-(tert-butoxy)-5-fluorobenzoic acid ≥95%
3-(tert-butoxy)-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29091336
CAS Number: 1369915-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR