4-Butyl-2-chloro-1-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

46839-500MG 46839-1G 46839-5G 46839-25G
77327-250EA 201.88 USD
77327-250 77350-826 77363-552 77386-132
4-Butyl-2-chloro-1-fluorobenzene ≥95%
4-Butyl-2-chloro-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClF
Storage Temperature: Ambient
MDL Number: MFCD32707206
CAS Number: 2432848-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR