2-Chloro-1-fluoro-4-pentylbenzene ≥97%

Supplier: AOBChem USA

Warning

46838-500MG 46838-1G 46838-5G 46838-25G
77327-248EA 202.91 USD
77327-248 77350-824 77363-550 77386-130
2-Chloro-1-fluoro-4-pentylbenzene ≥97%
2-Chloro-1-fluoro-4-pentylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClF
Storage Temperature: Ambient
MDL Number: MFCD32707205
CAS Number: 2432848-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR