2-Chloro-1-fluoro-4-isopentylbenzene ≥95%

Supplier: AOBChem USA

Warning

46837-500MG 46837-1G 46837-5G 46837-25G
77327-246EA 243.08 USD
77327-246 77350-822 77363-548 77386-128
2-Chloro-1-fluoro-4-isopentylbenzene ≥95%
2-Chloro-1-fluoro-4-isopentylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClF
Storage Temperature: Ambient
MDL Number: MFCD32707204
CAS Number: 2432849-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR