4-Butyl-1,2-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

46834-500MG 46834-1G 46834-5G
77335-804EA 366.68 USD
77335-804 77350-818 77375-098
4-Butyl-1,2-difluorobenzene ≥95%
4-Butyl-1,2-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂
MW: 170.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707202
CAS Number: 2432848-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR