1,2-Difluoro-4-propylbenzene ≥95%

Supplier: AOBChem USA

Warning

46833-500MG 46833-1G 46833-5G
77337-078EA 407.88 USD
77337-078 77350-816 77375-096
1,2-Difluoro-4-propylbenzene ≥95%
1,2-Difluoro-4-propylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂
MW: 156.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707201
CAS Number: 1397288-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR