2-Bromo-1-fluoro-4-pentylbenzene ≥95%

Supplier: AOBChem USA

Warning

46831-500MG 46831-1G 46831-5G 46831-25G
77327-244EA 239.99 USD
77327-244 77350-812 77363-546 77386-126
2-Bromo-1-fluoro-4-pentylbenzene ≥95%
2-Bromo-1-fluoro-4-pentylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrF
MW: 245.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707199
CAS Number: 2432848-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR