2-Bromo-4-butyl-1-fluorobenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
46830-500MG
46830-1G
46830-5G
77336-372EA
387.28
USD
77336-372
77350-810
77375-092
2-Bromo-4-butyl-1-fluorobenzene ≥95%
2-Bromo-4-butyl-1-fluorobenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₂BrF Storage Temperature: Ambient |
MDL Number:
MFCD32707198 CAS Number: 1632406-80-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...