N'-(4-fluoro-3-methoxybenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46821-500MG 46821-1G 46821-5G 46821-25G
77327-242EA 128.75 USD
77327-242 77344-244 77363-544 77375-088
N'-(4-fluoro-3-methoxybenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-fluoro-3-methoxybenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅FN₂O₃S
MW: 322.355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707197
CAS Number: 2414426-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR