1-Bromo-3-chloro-5-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

46798-500MG 46798-1G 46798-5G 46798-25G
77327-238EA 202.91 USD
77327-238 77350-804 77363-540 77386-122
1-Bromo-3-chloro-5-ethylbenzene ≥95%
1-Bromo-3-chloro-5-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MW: 219.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356415
CAS Number: 1369900-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR