(2-Methoxy-3,5-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

46733-500MG 46733-1G 46733-5G 46733-25G
77327-232EA 215.27 USD
77327-232 77350-796 77363-534 77386-120
(2-Methoxy-3,5-dimethylphenyl)methanol ≥97%
(2-Methoxy-3,5-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD21909892
CAS Number: 27326-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR