5-Chloro-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46706-500MG 46706-1G 46706-5G 46706-25G
77327-228EA 108.15 USD
77327-228 77343-174 77363-530 77375-072
5-Chloro-2-isopropoxybenzoic acid ≥97%
5-Chloro-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02729206
CAS Number: 634154-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR