5-Acetyl-3-bromo-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46704-500MG 46704-1G 46704-5G 46704-25G
77327-226EA 187.46 USD
77327-226 77350-794 77363-528 77386-118
5-Acetyl-3-bromo-2-fluorobenzoic acid ≥97%
5-Acetyl-3-bromo-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₆BrFO₃
MW: 261.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564102
CAS Number: 2734774-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR