1-(3-Bromo-4-fluoro-5-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

46703-500MG 46703-1G 46703-5G 46703-25G
77327-224EA 135.96 USD
77327-224 77344-510 77363-526 77386-116
1-(3-Bromo-4-fluoro-5-(methylthio)phenyl)ethanone ≥97%
1-(3-Bromo-4-fluoro-5-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈BrFOS
MW: 263.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756305
CAS Number: 2755718-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR