1-(3-Bromo-4-fluoro-5-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

46702-500MG 46702-1G 46702-5G 46702-25G
77327-222EA 236.9 USD
77327-222 77350-792 77363-524 77386-114
1-(3-Bromo-4-fluoro-5-methylphenyl)ethanone ≥97%
1-(3-Bromo-4-fluoro-5-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈BrFO
MW: 231.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31731075
CAS Number: 1782474-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR