(5-Bromopyridin-2-yl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

46659-500MG 46659-1G 46659-5G
77327-216EA 332.69 USD
77327-216 77350-786 77375-066
(5-Bromopyridin-2-yl)(thiomorpholino)methanone ≥97%
(5-Bromopyridin-2-yl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrN₂OS
MW: 287.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17259483
CAS Number: 1410447-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR