5-Bromo-N-isobutylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

46655-500MG 46655-1G 46655-5G 46655-25G
77327-214EA 181.28 USD
77327-214 77346-842 77363-518 77386-110
5-Bromo-N-isobutylpicolinamide ≥97%
5-Bromo-N-isobutylpicolinamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD17259451
CAS Number: 1339520-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR