Isopropyl 5-bromopicolinate ≥97%

Supplier: AOBChem USA

Warning

46652-500MG 46652-1G 46652-5G 46652-25G
77327-212EA 202.91 USD
77327-212 77350-784 77363-516 77386-108
Isopropyl 5-bromopicolinate ≥97%
Isopropyl 5-bromopicolinate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD21104740
CAS Number: 1289268-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR