Butyl 5-bromopicolinate ≥97%

Supplier: AOBChem USA

Warning

46650-500MG 46650-1G 46650-5G 46650-25G
77327-210EA 202.91 USD
77327-210 77350-782 77363-514 77386-106
Butyl 5-bromopicolinate ≥97%
Butyl 5-bromopicolinate

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD23721401
CAS Number: 1289092-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR