1,2-Dibromo-5-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

46643-500MG 46643-1G 46643-5G 46643-25G
77327-208EA 214.24 USD
77327-208 77350-780 77363-512 77386-104
1,2-Dibromo-5-fluoro-3-methoxybenzene ≥97%
1,2-Dibromo-5-fluoro-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂FO
MW: 283.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784576
CAS Number: 1805122-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR