N-Cyclopropyl-2-fluoronicotinamide ≥97%

Supplier: AOBChem USA

Warning

46605-500MG 46605-1G 46605-5G
77336-854EA 395.52 USD
77336-854 77350-776 77375-062
N-Cyclopropyl-2-fluoronicotinamide ≥97%
N-Cyclopropyl-2-fluoronicotinamide

Precaución: for Research Use Only

Formula: C₉H₉FN₂O
MW: 180.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32692990
CAS Number: 1513160-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR