N'-(2-Bromobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46559-500MG 46559-1G 46559-5G 46559-25G
77327-204EA 148.32 USD
77327-204 77345-280 77363-502 77386-100
N'-(2-Bromobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2-Bromobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrN₂O₂S
MW: 353.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00996847
CAS Number: 158918-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR