N'-(2-Chloro-6-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46558-500MG 46558-1G 46558-5G 46558-25G
77327-202EA 120.51 USD
77327-202 77343-712 77363-500 77375-048
N'-(2-Chloro-6-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2-Chloro-6-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFN₂O₂S
MW: 326.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01114439
CAS Number: 292077-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR