N'-(2-Bromo-6-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46554-500MG 46554-1G 46554-5G 46554-25G
77327-200EA 106.09 USD
77327-200 77343-124 77363-496 77375-044
N'-(2-Bromo-6-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2-Bromo-6-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707191
CAS Number: 2082699-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR