N'-(3-Bromo-4-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46552-1G 46552-5G 46552-25G
77341-144EA 72.1 USD
77341-144 77363-492 77375-040
N'-(3-Bromo-4-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Bromo-4-chlorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClN₂O₂S
MW: 387.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707190
CAS Number: 2432854-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR