N'-(4-Bromo-3-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46551-1G 46551-5G 46551-25G
77341-272EA 80.34 USD
77341-272 77363-490 77375-038
N'-(4-Bromo-3-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Bromo-3-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707189
CAS Number: 2416046-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR