N'-(4-Bromo-3-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46550-1G 46550-5G 46550-25G
77341-142EA 72.1 USD
77341-142 77363-488 77375-036
N'-(4-Bromo-3-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Bromo-3-chlorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClN₂O₂S
MW: 387.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707188
CAS Number: 2432855-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR