N'-(4-Chloro-3-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46549-500MG 46549-1G 46549-5G 46549-25G
77327-198EA 122.57 USD
77327-198 77343-910 77363-486 77375-034
N'-(4-Chloro-3-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Chloro-3-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFN₂O₂S
MW: 326.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707187
CAS Number: 2306459-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR