N'-(3,4-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46548-500MG 46548-1G 46548-5G 46548-25G
77327-196EA 107.12 USD
77327-196 77343-150 77363-484 77375-032
N'-(3,4-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3,4-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂N₂O₂S
MW: 310.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10182994
CAS Number: 457607-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR