N'-(3-Bromo-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥95%

Supplier: AOBChem USA

Warning

46547-500MG 46547-1G 46547-5G 46547-25G
77327-194EA 151.41 USD
77327-194 77345-424 77363-482 77386-098
N'-(3-Bromo-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥95%
N'-(3-Bromo-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707186
CAS Number: 2131827-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR