N'-(3-Chloro-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46546-500MG 46546-1G 46546-5G 46546-25G
77327-192EA 151.41 USD
77327-192 77345-390 77363-480 77386-096
N'-(3-Chloro-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Chloro-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFN₂O₂S
MW: 326.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707185
CAS Number: 2131827-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR