N'-(4-Bromo-3-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46545-1G 46545-5G 46545-25G
77340-678EA 60.77 USD
77340-678 77363-478 77375-030
N'-(4-Bromo-3-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Bromo-3-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrN₂O₂S
MW: 367.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564100
CAS Number: 2734780-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR