N'-(4-Chloro-3-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥95%

Supplier: AOBChem USA

Warning

46544-1G 46544-5G 46544-25G
77341-238EA 78.28 USD
77341-238 77363-476 77375-028
N'-(4-Chloro-3-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥95%
N'-(4-Chloro-3-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClN₂O₂S
MW: 322.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707184
CAS Number: 2484905-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR