N'-(3-Bromo-5-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46541-1G 46541-5G 46541-25G
77341-140EA 72.1 USD
77341-140 77363-472 77375-024
N'-(3-Bromo-5-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Bromo-5-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707182
CAS Number: 2432855-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR