N'-(3-Chloro-5-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46540-1G 46540-5G 46540-25G
77341-138EA 72.1 USD
77341-138 77363-470 77375-022
N'-(3-Chloro-5-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Chloro-5-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFN₂O₂S
MW: 326.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707181
CAS Number: 2432855-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR