N'-(3-Bromo-5-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46539-1G 46539-5G 46539-25G
77340-998EA 66.95 USD
77340-998 77363-468 77375-020
N'-(3-Bromo-5-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Bromo-5-chlorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClN₂O₂S
MW: 387.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707180
CAS Number: 2432855-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR