N'-(4-Chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46537-500MG 46537-1G 46537-5G 46537-25G
77327-190EA 99.91 USD
77327-190 77342-792 77363-466 77375-018
N'-(4-Chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Chlorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClN₂O₂S
MW: 308.783 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115920
CAS Number: 19350-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR