N'-(3-Bromo-5-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46535-1G 46535-5G 46535-25G
77341-136EA 72.1 USD
77341-136 77363-464 77375-016
N'-(3-Bromo-5-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Bromo-5-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrN₂O₂S
MW: 367.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707179
CAS Number: 2432855-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR